Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Synthware
™
storage tube with high vacuum valve
Products
Product Category
tubes (7)
Manufacturer
Capacity (mL)
Diameter (mm)
OD (mm)
Color
Package Size
Sterility
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
SYNF580825
Synthware
™
storage tube with high vacuum valve,
capacity 25 mL, valve size 0-8 mm, tube O.D. × L 26 mm × 110 mm
Expand
View Pricing
SYNF588250
Synthware
™
storage tube with high vacuum valve,
capacity 250 mL, valve size 0-8 mm, tube O.D. × L 60 mm × 165 mm
Expand
View Pricing
SYNF580025
Synthware
™
storage tube with high vacuum valve,
25 mL, valve size 0-4 mm, tube O.D. × L 26 mm × 110 mm
Expand
View Pricing
SYNF580850
Synthware
™
storage tube with high vacuum valve,
capacity 50 mL, valve size 0-8 mm, tube O.D. × L 28 mm × 120 mm
Expand
View Pricing
SYNF580050
Synthware
™
storage tube with high vacuum valve,
capacity 50 mL, valve size 0-4 mm, tube O.D. × L 28 mm × 120 mm
Expand
View Pricing
SYNF580100
Synthware
™
storage tube with high vacuum valve,
capacity 100 mL, valve size 0-4 mm, tube O.D. × L 40 mm × 165 mm
Expand
View Pricing
SYNF588100
Synthware
™
storage tube with high vacuum valve,
capacity 100 mL, valve size 0-8 mm, tube O.D. × L 40 mm × 165 mm
Expand
View Pricing