Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Swiss Boy lab jacks
Products
Color
Manufacturer
Length (mm)
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
Z635316
Swiss Boy lab jacks,
load ≤20 kg, platform L × W 50 mm × 40 mm, Model 100
Expand
View Pricing
Z635324
Swiss Boy lab jacks,
load ≤30 kg (middle position), platform L × W 80 mm × 75 mm, Model 105
Expand
View Pricing
Z635332
Swiss Boy lab jacks,
load ≤50 kg (middle position), platform L × W 120 mm × 140 mm, Model 110
Expand
View Pricing
Z635340
Swiss Boy lab jacks,
load ≤80 kg (middle position), platform L × W 122 mm × 150 mm, Model 115
Expand
View Pricing
Z635537
Swiss Boy lab jacks,
load ≤50 kg (w/o plates, middle position), platform L × W 200 mm × 200 mm, Model 110
Expand
View Pricing
Z635359
Swiss Boy lab jacks,
load ≤90 kg (middle position), platform L × W 180 mm × 216 mm, Model 116
Expand
View Pricing
Z635383
Swiss Boy lab jacks,
load ≤90 kg (middle position), platform L × W 240 mm × 320 mm, Model 118
Expand
View Pricing
Z635545
Swiss Boy lab jacks,
load ≤80 kg (with two plates, middle position), platform L × W 200 mm × 200 mm, Model 115
Expand
View Pricing
Z635553
Swiss Boy lab jacks,
load ≤90 kg (with two plates, middle position), platform L × W 400 mm × 500 mm, Model 118
Expand
View Pricing