Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Superconducting composite wire alloy, NbTi+Cu
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Filter Products
Showing 1-17 of 17
Compare
Product Number
Description
Pricing
GF38890165
Superconducting composite wire alloy, NbTi+Cu,
0.5m wire, 0.4mm diameter, copper/superconductor ratio 3:1 by volume
Expand
View Pricing
GF34901434
Superconducting composite wire alloy, NbTi+Cu,
50m wire, 0.4mm diameter, copper/superconductor ratio 3:1 by volume
Expand
View Pricing
GF73533786
Superconducting composite wire alloy, NbTi+Cu,
10m wire, 0.4mm diameter, copper/superconductor ratio 1.56:1 by volume
Expand
View Pricing
GF56279212
Superconducting composite wire alloy, NbTi+Cu,
10m wire, 0.4mm diameter, copper/superconductor ratio 3:1 by volume
Expand
View Pricing
GF34937231
Superconducting composite wire alloy, NbTi+Cu,
200m wire, 0.4mm diameter, copper/superconductor ratio 3:1 by volume
Expand
View Pricing
GF15055051
Superconducting composite wire alloy, NbTi+Cu,
0.5m wire, 0.4mm diameter, copper/superconductor ratio 1.35:1 by volume
Expand
View Pricing
GF86188270
Superconducting composite wire alloy, NbTi+Cu,
5m wire, 0.4mm diameter, copper/superconductor ratio 1.56:1 by volume
Expand
View Pricing
GF94289507
Superconducting composite wire alloy, NbTi+Cu,
1m wire, 0.4mm diameter, copper/superconductor ratio 1.56:1 by volume
Expand
View Pricing
GF61270804
Superconducting composite wire alloy, NbTi+Cu,
1m wire, 0.4mm diameter, copper/superconductor ratio 3:1 by volume
Expand
View Pricing
GF58850231
Superconducting composite wire alloy, NbTi+Cu,
5m wire, 0.4mm diameter, copper/superconductor ratio 3:1 by volume
Expand
View Pricing
GF11312413
Superconducting composite wire alloy, NbTi+Cu,
10m wire, 0.4mm diameter, copper/superconductor ratio 1.35:1 by volume
Expand
View Pricing
GF65445284
Superconducting composite wire alloy, NbTi+Cu,
100m wire, 0.4mm diameter, copper/superconductor ratio 1.56:1 by volume
Expand
View Pricing
GF67154317
Superconducting composite wire alloy, NbTi+Cu,
0.5m wire, 0.4mm diameter, copper/superconductor ratio 1.56:1 by volume
Expand
View Pricing
GF14051483
Superconducting composite wire alloy, NbTi+Cu,
50m wire, 0.4mm diameter, copper/superconductor ratio 1.35:1 by volume
Expand
View Pricing
GF91723563
Superconducting composite wire alloy, NbTi+Cu,
50m wire, 0.4mm diameter, copper/superconductor ratio 1.56:1 by volume
Expand
View Pricing
GF62679493
Superconducting composite wire alloy, NbTi+Cu,
100m wire, 0.4mm diameter, copper/superconductor ratio 3:1 by volume
Expand
View Pricing
GF35290492
Superconducting composite wire alloy, NbTi+Cu,
5m wire, 0.4mm diameter, copper/superconductor ratio 1.35:1 by volume
Expand
View Pricing