Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Supelguard HPLC Guard Cartridge Holder
Products
Product Category
holders (3)
columns (1)
Brand
Supelco (3)
Housing Material
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
59660-U
Supelguard HPLC Guard Cartridge Holder,
Stand-alone, for use with Supelguard cartridges (2 cm L. x 2.1 to 4.0 mm I.D.)
Expand
View Pricing
55205
Supelguard HPLC Guard Cartridge Holder,
Direct-Connect, for use with Supelguard cartridge (2 cm L. x 3 to 4.6 mm I.D.)
Expand
View Pricing
21150AST
Supelguard HPLC Guard Cartridge Holder,
Stand-Alone (Swivel-type), for use with Supelguard cartridges (2 cm L. x 2 to 4.6 mm I.D.)
Expand
View Pricing
504262
Supelguard HPLC Guard Cartridge Holder,
Direct-Connect (Swivel-type), for use with Supelguard cartridges (2 cm L. x 2.1 mm I.D.)
Expand
View Pricing