Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Supelclean
™
ENVI-Carb
™
SPE Tube
Products
Product Category
tubes (7)
Brand
Supelco (7)
Product Line
Chemical Composition
Separation Technique
Volume (ml)
Quality Segment
Application
Package Size
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
57094
Supelclean
™
ENVI-Carb
™
SPE Tube,
bed wt. 500 mg, volume 6 mL, pk of 30
Expand
View Pricing
57088
Supelclean
™
ENVI-Carb
™
SPE Tube,
bed wt. 250 mg, volume 3 mL, pk of 54
Expand
View Pricing
57092
Supelclean
™
ENVI-Carb
™
SPE Tube,
bed wt. 250 mg, volume 6 mL, pk of 30
Expand
View Pricing
57109-U
Supelclean
™
ENVI-Carb
™
SPE Tube,
bed wt. 100 mg, volume 1 mL, pk of 108
Expand
View Pricing
57127-U
Supelclean
™
ENVI-Carb
™
SPE Tube,
bed wt. 1 g, volume 12 mL, pk of 20
Expand
View Pricing
57130
Supelclean
™
ENVI-Carb
™
SPE Tube,
bed wt. 10 g, volume 60 mL, pk of 16
Expand
View Pricing
57128
Supelclean
™
ENVI-Carb
™
SPE Tube,
bed wt. 2 g, volume 12 mL, pk of 20
Expand
View Pricing