Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sulfamic acid
Synonym(s):
Amidosulfonic acid
Linear Formula:
NH
2
SO
3
H
CAS Number:
5329-14-6
Molecular Weight:
97.09
EC Number:
226-218-8
Products
Product Category
inorganic acids (3)
organic acids (2)
Brand
Sigma-Aldrich (4)
Supelco (2)
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Special Grade
analytical standard (2)
ACS reagent (1)
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
383120
Sulfamic acid,
ACS reagent, 99.3%
Expand
View Pricing
242772
Sulfamic acid,
ReagentPlus
®
, ≥99%
Expand
View Pricing
242780
Sulfamic acid,
reagent grade, 98%
Expand
View Pricing
07401
Sulfamic acid,
≥99.5% (alkalimetric)
Expand
View Pricing
481505
Sulfamic acid,
99.999% trace metals basis
Expand
View Pricing
86040
Sulfamic acid,
analytical standard (for acidimetry), ACS reagent
Expand
View Pricing
11358
Sulfamic acid,
analytical standard
Expand
View Pricing