Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Storage Tank
Products
Product Category
tanks (9)
Brand
Milli-Q (9)
Capacity (L)
Volume (ml)
Compatibility
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
TANKPE030
Storage Tank,
30 L polyethylene storage tank, An optimally integrated storage solution for your pure (Type 2/3) water
Expand
View Pricing
TANKA050
Storage Tank,
50 L Milli-Q
®
IQ/IX/EQ storage tank, Stores and maintains the pure (Type 2 and Type 3) water quality
Expand
View Pricing
TANKA100
Storage Tank,
100 L Milli-Q
®
IQ/IX/EQ storage tank, Stores and maintains the pure (Type 2 or Type 3) water quality
Expand
View Pricing
TANKA025
Storage Tank,
25 L Milli-Q
®
IQ/IX/EQ storage tank, Stores and maintains the pure (Type 2 or Type 3) water quality
Expand
View Pricing
TANKPE100
Storage Tank,
100 L polyethylene storage tank, An optimally integrated storage solution for your pure (Type 2/3) water
Expand
View Pricing
TANKPE060
Storage Tank,
60 L polyethylene storage tank, An optimally integrated storage solution for your pure (Type 2/3) water
Expand
View Pricing
TANKAFS10
Storage Tank,
10 L polyethylene storage tank, For optimal storage of pure (Type 2) water
Expand
View Pricing
ZBITANK01
Storage Tank,
100 L underbench polyethylene storage tank, Protects stored pure (Type 2/3) water
Expand
View Pricing
TANKBI030
Storage Tank,
30 L underbench polyethylene storage tank, An optimally integrated storage solution for your pure (Type 2/3) water
Expand
View Pricing