Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Stainless Steel - AISI 430 alloy, Fe81Cr17MnSiCSP
Products
Manufacturer
Physical Form
Filter Products
Showing 1-15 of 15
Compare
Product Number
Description
Pricing
GF55194464
Stainless Steel - AISI 430 alloy, Fe81Cr17MnSiCSP,
foil, 150 x 150mm, 0.15mm thickness, annealed
Expand
View Pricing
GF39632812
Stainless Steel - AISI 430 alloy, Fe81Cr17MnSiCSP,
foil, 1m coil, 0.70mm thickness, annealed
Expand
View Pricing
GF38941623
Stainless Steel - AISI 430 alloy, Fe81Cr17MnSiCSP,
foil, 50 x 50mm, 0.70mm thickness, annealed
Expand
View Pricing
GF51576712
Stainless Steel - AISI 430 alloy, Fe81Cr17MnSiCSP,
foil, 100 x 100mm, 0.70mm thickness, annealed
Expand
View Pricing
GF15724379
Stainless Steel - AISI 430 alloy, Fe81Cr17MnSiCSP,
foil, 25 x 25mm, 0.70mm thickness, annealed
Expand
View Pricing
GF40215440
Stainless Steel - AISI 430 alloy, Fe81Cr17MnSiCSP,
foil, 100 x 100mm, 0.15mm thickness, annealed
Expand
View Pricing
GF93061492
Stainless Steel - AISI 430 alloy, Fe81Cr17MnSiCSP,
foil, 25 x 25mm, 0.15mm thickness, annealed
Expand
View Pricing
GF39992844
Stainless Steel - AISI 430 alloy, Fe81Cr17MnSiCSP,
foil, 150 x 150mm, 0.70mm thickness, annealed
Expand
View Pricing
GF60130932
Stainless Steel - AISI 430 alloy, Fe81Cr17MnSiCSP,
foil, 0.5m coil, 0.70mm thickness, annealed
Expand
View Pricing
GF02206482
Stainless Steel - AISI 430 alloy, Fe81Cr17MnSiCSP,
foil, 2m coil, 0.15mm thickness, annealed
Expand
View Pricing
GF72733439
Stainless Steel - AISI 430 alloy, Fe81Cr17MnSiCSP,
foil, 100 x 100mm, 2.0mm thickness, annealed
Expand
View Pricing
GF90747658
Stainless Steel - AISI 430 alloy, Fe81Cr17MnSiCSP,
foil, 50 x 50mm, 2.0mm thickness, annealed
Expand
View Pricing
GF22618403
Stainless Steel - AISI 430 alloy, Fe81Cr17MnSiCSP,
foil, 0.5m coil, 0.15mm thickness, annealed
Expand
View Pricing
GF06136826
Stainless Steel - AISI 430 alloy, Fe81Cr17MnSiCSP,
foil, 1m coil, 0.15mm thickness, annealed
Expand
View Pricing
GF89066739
Stainless Steel - AISI 430 alloy, Fe81Cr17MnSiCSP,
foil, 2m coil, 0.70mm thickness, annealed
Expand
View Pricing