Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Stainless Steel - AISI 420 alloy, Fe86.7Cr13.0C0.3
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Length (mm)
Filter Products
Showing 1-15 of 15
Compare
Product Number
Description
Pricing
GF57681759
Stainless Steel - AISI 420 alloy, Fe86.7Cr13.0C0.3,
sheet, 300 x 300mm, 1.5mm thickness
Expand
View Pricing
GF77425274
Stainless Steel - AISI 420 alloy, Fe86.7Cr13.0C0.3,
foil, 100 x 100mm, 0.075mm thickness, annealed, light tight (lt)
Expand
View Pricing
GF39597205
Stainless Steel - AISI 420 alloy, Fe86.7Cr13.0C0.3,
foil, 25 x 25mm, 0.075mm thickness, annealed, light tight (lt)
Expand
View Pricing
GF09145885
Stainless Steel - AISI 420 alloy, Fe86.7Cr13.0C0.3,
foil, 50 x 50mm, 0.075mm thickness, annealed, light tight (lt)
Expand
View Pricing
GF76565357
Stainless Steel - AISI 420 alloy, Fe86.7Cr13.0C0.3,
100mm rod, 9.5mm diameter
Expand
View Pricing
GF28607430
Stainless Steel - AISI 420 alloy, Fe86.7Cr13.0C0.3,
foil, 2m coil, 0.075mm thickness, annealed
Expand
View Pricing
GF10409105
Stainless Steel - AISI 420 alloy, Fe86.7Cr13.0C0.3,
500mm rod, 9.5mm diameter
Expand
View Pricing
GF28717050
Stainless Steel - AISI 420 alloy, Fe86.7Cr13.0C0.3,
foil, 0.2m coil, 0.075mm thickness, annealed
Expand
View Pricing
GF61020076
Stainless Steel - AISI 420 alloy, Fe86.7Cr13.0C0.3,
1m wire, 1.0mm diameter, as drawn
Expand
View Pricing
GF40096717
Stainless Steel - AISI 420 alloy, Fe86.7Cr13.0C0.3,
foil, 1m coil, 0.075mm thickness, annealed
Expand
View Pricing
GF86323686
Stainless Steel - AISI 420 alloy, Fe86.7Cr13.0C0.3,
foil, 0.5m coil, 0.075mm thickness, annealed
Expand
View Pricing
GF40661298
Stainless Steel - AISI 420 alloy, Fe86.7Cr13.0C0.3,
10m wire, 1.0mm diameter, as drawn
Expand
View Pricing
GF56100473
Stainless Steel - AISI 420 alloy, Fe86.7Cr13.0C0.3,
50m wire, 1.0mm diameter, as drawn
Expand
View Pricing
GF82124940
Stainless Steel - AISI 420 alloy, Fe86.7Cr13.0C0.3,
20m wire, 1.0mm diameter, as drawn
Expand
View Pricing
GF88886266
Stainless Steel - AISI 420 alloy, Fe86.7Cr13.0C0.3,
5m wire, 1.0mm diameter, as drawn
Expand
View Pricing