Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Stainless Steel - AISI 410 alloy, FeCr12.5
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Length (mm)
Particle Size (µm)
Filter Products
Showing 1-16 of 16
Compare
Product Number
Description
Pricing
GF64171694
Stainless Steel - AISI 410 alloy, FeCr12.5,
powder, 45micron max. particle size, atomized, 100g
Expand
View Pricing
GF51524018
Stainless Steel - AISI 410 alloy, FeCr12.5,
powder, 45micron max. particle size, atomized, 1000g
Expand
View Pricing
GF40501639
Stainless Steel - AISI 410 alloy, FeCr12.5,
powder, 45micron max. particle size, atomized, 2000g
Expand
View Pricing
GF08236133
Stainless Steel - AISI 410 alloy, FeCr12.5,
foil, 50 x 50mm, 0.5mm thickness
Expand
View Pricing
GF82812191
Stainless Steel - AISI 410 alloy, FeCr12.5,
powder, 45micron max. particle size, atomized, 500g
Expand
View Pricing
GF99049348
Stainless Steel - AISI 410 alloy, FeCr12.5,
20m wire, 1mm diameter, as drawn
Expand
View Pricing
GF21462498
Stainless Steel - AISI 410 alloy, FeCr12.5,
powder, 45micron max. particle size, atomized, 200g
Expand
View Pricing
GF14906505
Stainless Steel - AISI 410 alloy, FeCr12.5,
foil, 100 x 100mm, 0.5mm thickness
Expand
View Pricing
GF60017903
Stainless Steel - AISI 410 alloy, FeCr12.5,
100mm rod, 9.5mm diameter
Expand
View Pricing
GF46462013
Stainless Steel - AISI 410 alloy, FeCr12.5,
500mm rod, 9.5mm diameter
Expand
View Pricing
GF71619677
Stainless Steel - AISI 410 alloy, FeCr12.5,
foil, 200 x 200mm, 0.5mm thickness
Expand
View Pricing
GF42008240
Stainless Steel - AISI 410 alloy, FeCr12.5,
10m wire, 1mm diameter, as drawn
Expand
View Pricing
GF17629038
Stainless Steel - AISI 410 alloy, FeCr12.5,
1m wire, 1mm diameter, as drawn
Expand
View Pricing
GF70180714
Stainless Steel - AISI 410 alloy, FeCr12.5,
200mm rod, 9.5mm diameter
Expand
View Pricing
GF14476923
Stainless Steel - AISI 410 alloy, FeCr12.5,
50m wire, 1mm diameter, as drawn
Expand
View Pricing
GF23648148
Stainless Steel - AISI 410 alloy, FeCr12.5,
5m wire, 1mm diameter, as drawn
Expand
View Pricing