Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Stainless steel - AISI 321
Products
Manufacturer
Physical Form
Length (m)
Length (mm)
OD (mm)
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
GF28887351
Stainless steel - AISI 321,
tube, Fe/Cr18%/Ni 9%/Ti, outside diameter 10.0 mm, length 100 mm, wall thickness 2.0 mm
Expand
View Pricing
GF00198389
Stainless steel - AISI 321,
wire, Fe/Cr18%/Ni 9%/Ti, 0.5 mm diameter, length 20 m, temper annealed
Expand
View Pricing
GF03045811
Stainless steel - AISI 321,
foil, Fe/Cr18%/Ni 9%/Ti, thickness 0.25 mm, length 0.5 m, coil width 150 mm
Expand
View Pricing
GF87512924
Stainless steel - AISI 321,
wire, Fe/Cr18%/Ni 9%/Ti%, length 10 m, 0.5 mm diameter, temper annealed
Expand
View Pricing
GF35209571
Stainless steel - AISI 321,
foil, Fe/Cr18%/Ni 9%/Ti, thickness 0.25 mm, length 10 m, coil width 150 mm
Expand
View Pricing
GF19831057
Stainless steel - AISI 321,
wire, Fe/Cr18%/Ni 9%/Ti, 0.5 mm diameter, length 100 m, temper annealed
Expand
View Pricing
GF39238094
Stainless steel - AISI 321,
foil, Fe/Cr18%/Ni 9%/Ti, thickness 0.25 mm, length 1 m, coil width 150 mm
Expand
View Pricing
GF33648600
Stainless steel - AISI 321,
wire, Fe/Cr18%/Ni 9%/Ti, 0.5 mm diameter, length 200 m, temper annealed
Expand
View Pricing
GF66715274
Stainless steel - AISI 321,
foil, thickness 0.25 mm, Fe/Cr18%/Ni 9%/Ti%, length 5 m, coil width 150 mm
Expand
View Pricing