Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Stainless steel - AISI 316
Products
Manufacturer
Physical Form
Length (m)
Length (mm)
Filter Products
Showing 1-15 of 15
Compare
Product Number
Description
Pricing
GF47211870
Stainless steel - AISI 316,
mesh, Fe/Cr18%/Ni 10%/Mo 3%, nominal aperture 0.085 mm, size 300 × 300 mm, wire diameter 0.05 mm
Expand
View Pricing
GF38749102
Stainless steel - AISI 316,
mesh, Fe/Cr18%/Ni 10%/Mo 3%, nominal aperture 0.085 mm, size 100 × 100 mm, wire diameter 0.05 mm
Expand
View Pricing
GF30959141
Stainless steel - AISI 316,
foil, Fe/Cr18%/Ni 10%/Mo 3%, thickness 0.01 mm, size 200 × 200 mm
Expand
View Pricing
GF43209390
Stainless steel - AISI 316,
rod, Fe/Cr18%/Ni 10%/Mo 3%, 10.0 mm diameter, length 1000 mm, temper as drawn
Expand
View Pricing
GF64528904
Stainless steel - AISI 316,
foil, Fe/Cr18%/Ni10%/Mo 3%, thickness 1.0 mm, size 15 × 45 mm, temper annealed
Expand
View Pricing
GF22533128
Stainless steel - AISI 316,
rod, Fe/Cr18%/Ni 10%/Mo 3%, 10.0 mm diameter, length 100 mm, temper as drawn
Expand
View Pricing
GF48132558
Stainless steel - AISI 316,
rod, Fe/Cr18%/Ni 10%/Mo 3%, 10.0 mm diameter, length 500 mm, temper as drawn
Expand
View Pricing
GF09808106
Stainless steel - AISI 316,
foil, Fe/Cr18%/Ni 10%/Mo 3%, thickness 0.05 mm, length 5 m, coil width 50 mm
Expand
View Pricing
GF77247153
Stainless steel - AISI 316,
foil, thickness 0.05 mm, Fe/Cr18%/Ni10%/Mo 3%, length 2 m, coil width 50 mm
Expand
View Pricing
GF02200194
Stainless steel - AISI 316,
rod, Fe/Cr18%/Ni 10%/Mo 3%, 10.0 mm diameter, length 200 mm, temper as drawn
Expand
View Pricing
GF33524805
Stainless steel - AISI 316,
foil, Fe/Cr18%/Ni 10%/Mo 3%, thickness 0.05 mm, length 0.5 m, coil width 50 mm
Expand
View Pricing
GF58077591
Stainless steel - AISI 316,
foil, Fe/Cr18%/Ni 10%/Mo 3%, thickness 0.0075 mm, size 25 × 25 mm, temper as rolled
Expand
View Pricing
GF55003238
Stainless steel - AISI 316,
foil, Fe/Cr18%/Ni 10%/Mo 3%, thickness 0.05 mm, length 1 m, coil width 50 mm
Expand
View Pricing
GF00560720
Stainless steel - AISI 316,
foil, Fe/Cr18%/Ni 10%/Mo 3%, thickness 0.0075 mm, size 50 × 50 mm, temper as rolled
Expand
View Pricing
GF58391950
Stainless steel - AISI 316,
foil, Fe/Cr18%/Ni 10%/Mo 3%, thickness 0.05 mm, length 10 m, coil width 50 mm
Expand
View Pricing