Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Stainless steel - AISI 304L alloy, Fe/Cr18/Ni10
Products
Manufacturer
Physical Form
Diameter (mm)
Length (mm)
Filter Products
Showing 1-20 of 43
Page 1 of 3
Page 1 of 3
Compare
Product Number
Description
Pricing
GF18650123
Stainless steel - AISI 304L alloy, Fe/Cr18/Ni10,
foil, 300 x 300mm, 0.15mm thickness, annealed
Expand
View Pricing
GF57609883
Stainless steel - AISI 304L alloy, Fe/Cr18/Ni10,
foil, 2m coil, 0.3mm thickness, annealed
Expand
View Pricing
GF38255632
Stainless steel - AISI 304L alloy, Fe/Cr18/Ni10,
foil, 1m coil, 0.2mm thickness, annealed
Expand
View Pricing
GF88457686
Stainless steel - AISI 304L alloy, Fe/Cr18/Ni10,
foil, 50 x 50mm, 0.15mm thickness, annealed
Expand
View Pricing
GF93209991
Stainless steel - AISI 304L alloy, Fe/Cr18/Ni10,
200mm tube, 7.0mm outside diameter, 1mm wall thickness, 5.0mm inside diameter, annealed
Expand
View Pricing
GF73480989
Stainless steel - AISI 304L alloy, Fe/Cr18/Ni10,
100mm tube, 7.0mm outside diameter, 1mm wall thickness, 5.0mm inside diameter, annealed
Expand
View Pricing
GF18287365
Stainless steel - AISI 304L alloy, Fe/Cr18/Ni10,
foil, 100 x 100mm, 1.0mm thickness, annealed
Expand
View Pricing
GF98150340
Stainless steel - AISI 304L alloy, Fe/Cr18/Ni10,
foil, 150 x 150mm, 0.15mm thickness, annealed
Expand
View Pricing
GF65463756
Stainless steel - AISI 304L alloy, Fe/Cr18/Ni10,
1000mm wire, 0.3mm diameter, annealed, straight wire
Expand
View Pricing
GF84848316
Stainless steel - AISI 304L alloy, Fe/Cr18/Ni10,
foil, 100 x 100mm, 0.75mm thickness, annealed
Expand
View Pricing
GF19498360
Stainless steel - AISI 304L alloy, Fe/Cr18/Ni10,
foil, 150 x 150mm, 0.3mm thickness, annealed
Expand
View Pricing
GF25932809
Stainless steel - AISI 304L alloy, Fe/Cr18/Ni10,
200mm rod, 8.0mm diameter
Expand
View Pricing
GF62208660
Stainless steel - AISI 304L alloy, Fe/Cr18/Ni10,
100mm rod, 8.0mm diameter
Expand
View Pricing
GF38232367
Stainless steel - AISI 304L alloy, Fe/Cr18/Ni10,
500mm rod, 2mm diameter
Expand
View Pricing
GF13147949
Stainless steel - AISI 304L alloy, Fe/Cr18/Ni10,
foil, 300 x 300mm, 0.3mm thickness, annealed
Expand
View Pricing
GF73617925
Stainless steel - AISI 304L alloy, Fe/Cr18/Ni10,
100mm rod, 2mm diameter
Expand
View Pricing
GF42273266
Stainless steel - AISI 304L alloy, Fe/Cr18/Ni10,
foil, 100 x 200mm, 0.9mm thickness, annealed, mirror polished on one side
Expand
View Pricing
GF69141540
Stainless steel - AISI 304L alloy, Fe/Cr18/Ni10,
foil, 2m coil, 0.75mm thickness, annealed
Expand
View Pricing
GF54806722
Stainless steel - AISI 304L alloy, Fe/Cr18/Ni10,
foil, 50 x 50mm, 0.75mm thickness, annealed
Expand
View Pricing
GF52297195
Stainless steel - AISI 304L alloy, Fe/Cr18/Ni10,
foil, 1m coil, 0.3mm thickness, annealed
Expand
View Pricing
Page 1 of 3
Page 1 of 3