Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Stainless steel - AISI 304
Products
Manufacturer
Physical Form
Diameter (mm)
Length (mm)
Filter Products
Showing 1-20 of 33
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
GF22415699
Stainless steel - AISI 304,
mesh, Fe/Cr18%/Ni 10%, nominal aperture 0.54 mm, size 100 × 100 mm, thickness 0.6 mm
Expand
View Pricing
GF08769123
Stainless steel - AISI 304,
foil, Fe/Cr18%/Ni 10%, thickness 0.1 mm, size 300 × 300 mm, temper hard
Expand
View Pricing
GF99424312
Stainless steel - AISI 304,
mesh, size 0.54 mm, Fe/Cr18%/Ni10%, size 150 × 150 mm, thickness 0.6 mm
Expand
View Pricing
GF65267174
Stainless steel - AISI 304,
foil, thickness 0.1 mm, Fe/Cr18%/Ni10%, size 25 × 25 mm, temper hard
Expand
View Pricing
GF17261647
Stainless steel - AISI 304,
tube, Fe/Cr18%/Ni 10%, outside diameter 0.9 mm, length 35 mm, wall thickness 0.15 mm
Expand
View Pricing
GF82989444
Stainless steel - AISI 304,
foil, thickness 1.5 mm, Fe/Cr18%/Ni10%, size 100 × 100 mm
Expand
View Pricing
GF68936529
Stainless steel - AISI 304,
foil, thickness 0.1 mm, Fe/Cr18%/Ni10%, length 1 m, temper hard
Expand
View Pricing
GF34069091
Stainless steel - AISI 304,
foil, Fe/Cr18%/Ni 10%, thickness 0.1 mm, size 150 × 150 mm, temper hard
Expand
View Pricing
GF90669314
Stainless steel - AISI 304,
foil, Fe/Cr18%/Ni10%, thickness 0.1 mm, temper hard, size 50 × 50 mm
Expand
View Pricing
GF58021231
Stainless steel - AISI 304,
mesh, Fe/Cr18%/Ni 10%, nominal aperture 0.54 mm, size 600 × 600 mm, thickness 0.6 mm
Expand
View Pricing
GF50550285
Stainless steel - AISI 304,
mesh, Fe/Cr18%/Ni 10%, nominal aperture 0.54 mm, size 900 × 900 mm, thickness 0.6 mm
Expand
View Pricing
GF07910254
Stainless steel - AISI 304,
rod, Fe/Cr18%/Ni 10%, 10.0 mm diameter, length 200 mm, temper as drawn
Expand
View Pricing
GF48743635
Stainless steel - AISI 304,
mesh, Fe/Cr18%/Ni 10%, nominal aperture 0.54 mm, size 300 × 300 mm, thickness 0.6 mm
Expand
View Pricing
GF47818200
Stainless steel - AISI 304,
rod, Fe/Cr18%/Ni 10%, 5.0 mm diameter, length 500 mm, temper as drawn
Expand
View Pricing
GF62112181
Stainless steel - AISI 304,
foil, thickness 0.1 mm, Fe/Cr18%/Ni10%, diameter 10 mm, temper hard
Expand
View Pricing
GF69585722
Stainless steel - AISI 304,
rod, 10.0 mm diameter, Fe/Cr18%/Ni10%, length 100 mm, temper as drawn
Expand
View Pricing
GF16035173
Stainless steel - AISI 304,
foil, Fe/Cr18%/Ni 10%, thickness 0.1 mm, 15 mm diameter, temper hard
Expand
View Pricing
GF20156464
Stainless steel - AISI 304,
foil, Fe/Cr18%/Ni 10%, thickness 0.7 mm, size 100 × 100 mm, temper annealed
Expand
View Pricing
GF26621265
Stainless steel - AISI 304,
rod, Fe/Cr18%/Ni 10%, 10.0 mm diameter, length 1000 mm, temper as drawn
Expand
View Pricing
GF28666378
Stainless steel - AISI 304,
foil, Fe/Cr18%/Ni 10%, thickness 1.5 mm, size 150 × 150 mm
Expand
View Pricing
Page 1 of 2
Page 1 of 2