Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Stainless Steel - 15-7PH alloy, FeCr15Ni7Mo2.25
Products
Manufacturer
Physical Form
Length (m)
Filter Products
Showing 1-20 of 24
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
GF69843426
Stainless Steel - 15-7PH alloy, FeCr15Ni7Mo2.25,
foil, 100 x 100mm, 0.05mm thickness, annealed, precipitation hardenable, light tight (lt)
Expand
View Pricing
GF21401873
Stainless Steel - 15-7PH alloy, FeCr15Ni7Mo2.25,
foil, 6mm disks, 0.25mm thickness, annealed, precipitation hardenable
Expand
View Pricing
GF94247770
Stainless Steel - 15-7PH alloy, FeCr15Ni7Mo2.25,
foil, 50 x 50mm, 0.25mm thickness, annealed, precipitation hardenable
Expand
View Pricing
GF70158742
Stainless Steel - 15-7PH alloy, FeCr15Ni7Mo2.25,
foil, 50 x 50mm, 0.05mm thickness, annealed, precipitation hardenable, light tight (lt)
Expand
View Pricing
GF16171219
Stainless Steel - 15-7PH alloy, FeCr15Ni7Mo2.25,
foil, 4mm disks, 0.25mm thickness, annealed, precipitation hardenable
Expand
View Pricing
GF30428377
Stainless Steel - 15-7PH alloy, FeCr15Ni7Mo2.25,
foil, 150 x 150mm, 0.05mm thickness, annealed, precipitation hardenable, light tight (lt)
Expand
View Pricing
GF21421796
Stainless Steel - 15-7PH alloy, FeCr15Ni7Mo2.25,
foil, 150 x 150mm, 0.25mm thickness, annealed, precipitation hardenable
Expand
View Pricing
GF98353985
Stainless Steel - 15-7PH alloy, FeCr15Ni7Mo2.25,
foil, 25 x 25mm, 0.25mm thickness, annealed, precipitation hardenable
Expand
View Pricing
GF98961261
Stainless Steel - 15-7PH alloy, FeCr15Ni7Mo2.25,
foil, 25 x 25mm, 0.05mm thickness, annealed, precipitation hardenable, light tight (lt)
Expand
View Pricing
GF46133456
Stainless Steel - 15-7PH alloy, FeCr15Ni7Mo2.25,
foil, 15mm disks, 0.25mm thickness, annealed, precipitation hardenable
Expand
View Pricing
GF62598707
Stainless Steel - 15-7PH alloy, FeCr15Ni7Mo2.25,
foil, 25mm disks, 0.05mm thickness, annealed, precipitation hardenable
Expand
View Pricing
GF17418405
Stainless Steel - 15-7PH alloy, FeCr15Ni7Mo2.25,
foil, 10mm disks, 0.05mm thickness, annealed, precipitation hardenable
Expand
View Pricing
GF24838713
Stainless Steel - 15-7PH alloy, FeCr15Ni7Mo2.25,
foil, 50mm disks, 0.05mm thickness, annealed, precipitation hardenable
Expand
View Pricing
GF03546277
Stainless Steel - 15-7PH alloy, FeCr15Ni7Mo2.25,
foil, 300 x 300mm, 0.05mm thickness, annealed, precipitation hardenable, light tight (lt)
Expand
View Pricing
GF15010928
Stainless Steel - 15-7PH alloy, FeCr15Ni7Mo2.25,
foil, 4mm disks, 0.05mm thickness, annealed, precipitation hardenable
Expand
View Pricing
GF43284168
Stainless Steel - 15-7PH alloy, FeCr15Ni7Mo2.25,
foil, 1m coil, 0.05mm thickness, annealed, precipitation hardenable
Expand
View Pricing
GF66681636
Stainless Steel - 15-7PH alloy, FeCr15Ni7Mo2.25,
foil, 25mm disks, 0.25mm thickness, annealed, precipitation hardenable
Expand
View Pricing
GF04369068
Stainless Steel - 15-7PH alloy, FeCr15Ni7Mo2.25,
foil, 10mm disks, 0.25mm thickness, annealed, precipitation hardenable
Expand
View Pricing
GF45739345
Stainless Steel - 15-7PH alloy, FeCr15Ni7Mo2.25,
foil, 8mm disks, 0.05mm thickness, annealed, precipitation hardenable
Expand
View Pricing
GF10520086
Stainless Steel - 15-7PH alloy, FeCr15Ni7Mo2.25,
foil, 2m coil, 0.05mm thickness, annealed, precipitation hardenable
Expand
View Pricing
Page 1 of 2
Page 1 of 2