Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Stainless Steel
Products
Manufacturer
Format
Quality Segment
Reference Material Type
Special Grade
certified reference material (15)
Technique
Filter Products
Showing 1-15 of 15
Compare
Product Number
Description
Pricing
NIST1155A
Stainless Steel,
NIST
®
SRM
®
1155a, Cr 18 - Ni 12 - Mo 2 (AISI 316)
Expand
View Pricing
NISTC1296
Stainless Steel,
NIST
®
SRM
®
C1296
Expand
View Pricing
NIST166C
Stainless Steel,
NIST
®
SRM
®
166c, low-carbon (AISI 316L)
Expand
View Pricing
NIST160B
Stainless Steel,
NIST
®
SRM
®
160b, Cr 18 - Ni 12 - Mo 2 (AISI 316)
Expand
View Pricing
NISTC1154A
Stainless Steel,
NIST
®
SRM
®
C1154a, 19 Cr - 13 Ni
Expand
View Pricing
NIST123C
Stainless Steel,
NIST
®
SRM
®
123c, Cr 17 - Ni 11 - Nb 0.6 (AISI 348)
Expand
View Pricing
NIST895
Stainless Steel,
NIST
®
SRM
®
895, (SAE 201)
Expand
View Pricing
NIST1297
Stainless Steel,
NIST
®
SRM
®
1297, (SAE 201)
Expand
View Pricing
NIST1172
Stainless Steel,
NIST
®
SRM
®
1172, Cr 17 - Ni 11 - Nb 0.6 (AISI 348)
Expand
View Pricing
NIST1219
Stainless Steel,
NIST
®
SRM
®
1219, Cr 16 - Ni 2 (AISI 431)
Expand
View Pricing
NIST1171
Stainless Steel,
NIST
®
SRM
®
1171, Cr 17 - Ni 11 - Ti 0.3 (AISI 321)
Expand
View Pricing
NIST893
Stainless Steel,
NIST
®
SRM
®
893, (SAE 405)
Expand
View Pricing
NIST1295
Stainless Steel,
NIST
®
SRM
®
1295, (SAE 405)
Expand
View Pricing
NISTC1151A
Stainless Steel,
NIST
®
SRM
®
C1151a, 23 Cr - 7 Ni
Expand
View Pricing
NISTC1153A
Stainless Steel,
NIST
®
SRM
®
C1153a, 17 Cr - 9 Ni
Expand
View Pricing