Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Spilfyter
®
hazmat spill control, polypropylene
Products
Brand
Sigma-Aldrich (5)
Capacity (L)
Capacity (mL)
Length (in)
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
Z158046
Spilfyter
®
hazmat spill control, polypropylene,
Pillow, capacity 1 L
Expand
View Pricing
Z158054
Spilfyter
®
hazmat spill control, polypropylene,
Pillow, capacity 4 L
Expand
View Pricing
Z201057
Spilfyter
®
hazmat spill control, polypropylene,
Pad, W × L 12 in. × 12 in.
Expand
View Pricing
Z201030
Spilfyter
®
hazmat spill control, polypropylene,
Roll, W × L 16 in. × 150 ft
Expand
View Pricing
Z550833
Spilfyter
®
hazmat spill control, polypropylene,
Pad, W × L 16 in. × 18 in., double weight
Expand
View Pricing
Z201073
Spilfyter
®
hazmat spill control, polypropylene,
Sock, W × L 3 in. × 4 ft
Expand
View Pricing
Z158038
Spilfyter
®
hazmat spill control, polypropylene,
Pillow, capacity 250 mL
Expand
View Pricing