Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Spermidine
Synonym(s):
1,8-Diamino-4-azaoctane,
N
-(3-Aminopropyl)-1,4-diaminobutane, Spermidine
Linear Formula:
NH
2
(CH
2
)
3
NH(CH
2
)
4
NH
2
CAS Number:
124-20-9
Molecular Weight:
145.25
EC Number:
204-689-0
Products
Product Category
bases (1)
peptides (1)
stabilizers (1)
Brand
SAFC (1)
Sigma-Aldrich (3)
Supelco (1)
Biological Source
Formula Weight
Gene Alias
pH Value
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
GMO free (1)
GMP (1)
analytical standard (1)
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
S0266
Spermidine,
BioReagent, for molecular biology, suitable for cell culture, ≥98%
Expand
View Pricing
S2626
Spermidine,
≥99% (GC)
Expand
View Pricing
85558
Spermidine,
BioUltra, for molecular biology, ≥99.5% (GC)
Expand
View Pricing
1.37165
Spermidine,
EMPROVE
®
EXPERT
Expand
View Pricing
49761
Spermidine,
analytical standard
Expand
View Pricing