Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Spectra-Por
®
Float-A-Lyzer
®
G2
Products
Manufacturer
Color
Package Size
MWCO (kDa)
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
Z727040
Spectra-Por
®
Float-A-Lyzer
®
G2,
blue, 1 mL, MWCO 100 kDa
Expand
View Pricing
Z727253
Spectra-Por
®
Float-A-Lyzer
®
G2,
blue, 10 mL, MWCO 100 kDa
Expand
View Pricing
Z727156
Spectra-Por
®
Float-A-Lyzer
®
G2,
blue, 5 mL, MWCO 100 kDa
Expand
View Pricing
Z726060
Spectra-Por
®
Float-A-Lyzer
®
G2,
black, 1 mL, MWCO 3.5-5 kDa
Expand
View Pricing
Z726710
Spectra-Por
®
Float-A-Lyzer
®
G2,
red, 1 mL, MWCO 20 kDa
Expand
View Pricing
Z726605
Spectra-Por
®
Float-A-Lyzer
®
G2,
yellow, 10 mL, MWCO 8-10 kDa
Expand
View Pricing
Z726273
Spectra-Por
®
Float-A-Lyzer
®
G2,
black, 10 mL, MWCO 3.5-5 kDa
Expand
View Pricing
Z726508
Spectra-Por
®
Float-A-Lyzer
®
G2,
yellow, 5 mL, MWCO 8-10 kDa
Expand
View Pricing
Z726834
Spectra-Por
®
Float-A-Lyzer
®
G2,
red, 5 mL, MWCO 20 kDa
Expand
View Pricing
Z726176
Spectra-Por
®
Float-A-Lyzer
®
G2,
black, 5 mL, MWCO 3.5-5 kDa
Expand
View Pricing
Z726931
Spectra-Por
®
Float-A-Lyzer
®
G2,
red, 10 mL, MWCO 20 kDa
Expand
View Pricing