Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
SP Bel-Art
®
Sterileware
™
Long Handled Sampling Spoon
Products
Product Category
spatulas/scoops (12)
Manufacturer
Length (cm)
Length (in)
Color
Package Size
Sterility
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
BAH369460000
SP Bel-Art
®
Sterileware
™
Long Handled Sampling Spoon,
white polystyrene, nominal capacity 14.79 mL , 3tsp, L × W 17.8 cm (7 in.) × 1.5625 in. (40 mm) mL, sterile, pack of 200 ea (individually wrapped)
Expand
View Pricing
BAH369460010
SP Bel-Art
®
Sterileware
™
Long Handled Sampling Spoon,
white polystyrene, nominal capacity 14.79 mL , 3tsp, L × W 17.8 cm (7 in.) × 1.5625 in. (40 mm), sterile, pack of 10 ea (individually wrapped)
Expand
View Pricing
BAH369400000
SP Bel-Art
®
Sterileware
™
Long Handled Sampling Spoon,
white polystyrene, nominal capacity 1.25 mL , .25¼tsp, L × W 7 in. (17.8 cm) × 0.8125 in. (20 mm), sterile, pack of 200 ea (individually wrapped)
Expand
View Pricing
BAH369420000
SP Bel-Art
®
Sterileware
™
Long Handled Sampling Spoon,
white polystyrene, nominal capacity 2.46 mL , ½tsp, L × W 17.8 cm (7 in.) × 1 in. (25 mm), sterile, pack of 200 ea (individually wrapped)
Expand
View Pricing
BAH369420010
SP Bel-Art
®
Sterileware
™
Long Handled Sampling Spoon,
white polystyrene, nominal capacity 2.46 mL , ½tsp, L × W 7 in. (178 mm) × 1 in. (25 mm), sterile, pack of 10 ea (individually wrapped)
Expand
View Pricing
BAH369440000
SP Bel-Art
®
Sterileware
™
Long Handled Sampling Spoon,
white polystyrene, nominal capacity 4.93 mL , 1 tsp, L × W 17.8 cm (7 in.) × 1.1875 in. (30 mm), sterile, pkg of 200 ea (individually wrapped)
Expand
View Pricing
BAH369440010
SP Bel-Art
®
Sterileware
™
Long Handled Sampling Spoon,
white polystyrene, nominal capacity 4.93 mL , 1 tsp, L × W 17.8 cm (7 in.) × 1.1875 in. (30 mm), sterile, pack of 10 ea (individually wrapped)
Expand
View Pricing
BAH369400010
SP Bel-Art
®
Sterileware
™
Long Handled Sampling Spoon,
white polystyrene, nominal capacity 1.25 mL , ¼tsp, L × W 7 in. (17.8 cm) × 0.8125 in. (20 mm), sterile, pack of 10 ea (individually wrapped)
Expand
View Pricing
BAH369421010
SP Bel-Art
®
Sterileware
™
Long Handled Sampling Spoon,
white polystyrene, nominal capacity 2.46 mL , 1/2tsp, L × W 17.8 cm (7 in.) × 1 in. (25 mm), sterile, pack of 10 ea (double wrapped)
Expand
View Pricing
BAH369461010
SP Bel-Art
®
Sterileware
™
Long Handled Sampling Spoon,
white polystyrene, nominal capacity 14.79 mL , 3tsp, L × W 17.8 cm (7 in.) × 1.5625 in. (40 mm), sterile, pkg of 10 ea (individually wrapped)
Expand
View Pricing
BAH369441010
SP Bel-Art
®
Sterileware
™
Long Handled Sampling Spoon,
white polystyrene, nominal capacity 4.93 mL , 1tsp, L × W 17.8 cm (7 in.) × 1.1875 in. (30 mm), sterile, pack of 10 ea (double wrapped)
Expand
View Pricing
BAH369401010
SP Bel-Art
®
Sterileware
™
Long Handled Sampling Spoon,
white polystyrene, nominal capacity 1.25 mL , ¼tsp, L × W 17.8 cm (7 in.) × 0.8125 in. (20 mm), sterile, pack of 10 ea (double bagged)
Expand
View Pricing