Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
SP Bel-Art
®
Spinbar
®
magnetic stirring fleas
Synonym(s):
Spinbar
®
magnetic stirring fleas
Products
Product Category
stir bars (3)
Manufacturer
Diameter (mm)
Length (in)
Color
Package Size
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
BAF371190083
SP Bel-Art
®
Spinbar
®
magnetic stirring fleas,
white, L × diam. 0.315 in. (8 mm) × 0.118 in. (3 mm)
Expand
View Pricing
BAF371210019
SP Bel-Art
®
Spinbar
®
magnetic stirring fleas,
red, L × diam. 0.25 in. (6.35 mm) × 0.118 in. (3 mm)
Expand
View Pricing
BAF371210027
SP Bel-Art
®
Spinbar
®
magnetic stirring fleas,
blue, L × diam. 0.5 in. (12.7 mm) × 0.118 in. (3 mm)
Expand
View Pricing