Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sorenson
™
low binding aerosol barrier tips
Products
Product Category
pipette tips (9)
Manufacturer
Binding Type
Color
Package Size
Sterility
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
Z719390
Sorenson
™
low binding aerosol barrier tips,
MicroReach Guard, maximum volume 10 μL
Expand
View Pricing
Z719447
Sorenson
™
low binding aerosol barrier tips,
MultiGuard, maximum volume 200 μL , Graduated
Expand
View Pricing
Z719463
Sorenson
™
low binding aerosol barrier tips,
MultiGuard, maximum volume 1000 μL
Expand
View Pricing
Z719412
Sorenson
™
low binding aerosol barrier tips,
MultiGuard, maximum volume 20 μL
Expand
View Pricing
Z719420
Sorenson
™
low binding aerosol barrier tips,
MultiGuard, maximum volume 100 μL
Expand
View Pricing
Z719439
Sorenson
™
low binding aerosol barrier tips,
MultiGuard, no graduation, maximum volume 200 μL
Expand
View Pricing
Z719404
Sorenson
™
low binding aerosol barrier tips,
MultiGuard E, maximum volume 10 μL
Expand
View Pricing
Z719374
Sorenson
™
low binding aerosol barrier tips,
MicroGuard G, maximum volume 10 μL
Expand
View Pricing
Z719471
Sorenson
™
low binding aerosol barrier tips,
MultiGuard, with graduation lines, maximum volume 200 μL
Expand
View Pricing