
Sodium trifluoroacetate

Synonym(s):
Trifluoroacetic acid sodium salt, Sodium trifluoroacetate
Linear Formula:
CF3COONa
CAS Number:
Molecular Weight:
136.01
Products
Product Category
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Format
Functional Group
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing