Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium tetraborate decahydrate
Synonym(s):
Borax decahydrate, Sodium borate decahydrate
Linear Formula:
Na
2
B
4
O
7
· 10H
2
O
CAS Number:
1303-96-4
Molecular Weight:
381.37
EC Number:
215-540-4
Products
Product Category
salts (3)
buffers (1)
Brand
Sigma-Aldrich (4)
Feature
Formula Weight
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Special Grade
ACS reagent (2)
Puriss (1)
Puriss P.A. (1)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
S9640
Sodium tetraborate decahydrate,
ACS reagent, ≥99.5%
Expand
View Pricing
B9876
Sodium tetraborate decahydrate,
ReagentPlus
®
, ≥99.5%
Expand
View Pricing
31457
Sodium tetraborate decahydrate,
puriss. p.a., ACS reagent, reag. ISO, buffer substance, ≥99.5%
Expand
View Pricing
B3545
Sodium tetraborate decahydrate,
BioXtra, ≥99.5%
Expand
View Pricing