Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium perchlorate monohydrate
Linear Formula:
NaClO
4
· H
2
O
CAS Number:
7791-07-3
Molecular Weight:
140.46
EC Number:
231-511-9
Products
Product Category
salts (2)
buffers (1)
Brand
Sigma-Aldrich (2)
Supelco (1)
Formula Weight
pH Value
Physical Form
Purity
Reagent Type
Special Grade
ACS reagent (2)
Puriss (1)
Puriss P.A. (1)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
310514
Sodium perchlorate monohydrate,
ACS reagent
Expand
View Pricing
56581
Sodium perchlorate monohydrate,
suitable for HPLC, LiChropur
™
, 99.0-101.0% (T)
Expand
View Pricing
71853-M
Sodium perchlorate monohydrate,
puriss. p.a., ACS reagent, ≥98.0% (T)
Expand
View Pricing