Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium methoxide solution
Synonym(s):
Sodium methylate
Linear Formula:
CH
3
ONa
CAS Number:
124-41-4
Molecular Weight:
54.02
Products
Product Category
organic bases (1)
Brand
Sigma-Aldrich (2)
Supelco (1)
Boiling Point (°C)
Formula Weight
Physical Form
Quality Segment
Special Grade
ACS reagent (1)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
156256
Sodium methoxide solution,
25 wt. % in methanol
Expand
View Pricing
403067
Sodium methoxide solution,
ACS reagent, 0.5 M CH
3
ONa in methanol (0.5N)
Expand
View Pricing
92446
Sodium methoxide solution,
0.5 M, for GC derivatization
Expand
View Pricing