Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
SmartPor, alumina membrane
Linear Formula:
Al
Products
Product Category
membranes (27)
Brand
Sigma-Aldrich (27)
Filter Products
Showing 1-20 of 27
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
SM75501010
SmartPor, alumina membrane,
thickness 50 μm, pore diameter 75 nm, pore size 125 nm (interpore distance), size 10 mm × 10 mm, pkg of 10 pieces
Expand
View Pricing
SM25501010
SmartPor, alumina membrane,
thickness 50 μm, pore diameter 25 nm, pore size 65 nm (interpore distance), size 10 mm × 10 mm, pkg of 10 pieces
Expand
View Pricing
SM40501010
SmartPor, alumina membrane,
thickness 50 μm, pore diameter 40 nm, pore size 125 nm (interpore distance), size 10 mm × 10 mm, pkg of 10 pieces
Expand
View Pricing
SM4050201
SmartPor, alumina membrane,
thickness 50 μm, pore diameter 40 nm, pore size 125 nm (interpore distance), size 20 mm × 20 mm, pkg of 1 pieces
Expand
View Pricing
SM60501010
SmartPor, alumina membrane,
thickness 50 μm, pore diameter 60 nm, pore size 125 nm (interpore distance), size 10 mm × 10 mm, pkg of 10 pieces
Expand
View Pricing
SM251001010
SmartPor, alumina membrane,
thickness 100 μm, pore diameter 25 nm, pore size 65 nm (interpore distance), size 10 mm × 10 mm, pkg of 10 pieces
Expand
View Pricing
SM2550205
SmartPor, alumina membrane,
thickness 50 μm, pore diameter 25 nm, pore size 65 nm (interpore distance), size 20 mm × 20 mm, pkg of 5 pieces
Expand
View Pricing
SM4050105
SmartPor, alumina membrane,
thickness 50 μm, pore diameter 40 nm, pore size 125 nm (interpore distance), size 10 mm × 10 mm, pkg of 5 pieces
Expand
View Pricing
SM6050105
SmartPor, alumina membrane,
thickness 50 μm, pore diameter 60 nm, pore size 125 nm (interpore distance), size 10 mm × 10 mm, pkg of 5 pieces
Expand
View Pricing
SM25100205
SmartPor, alumina membrane,
thickness 100 μm, pore diameter 25 nm, pore size 65 nm (interpore distance), size 20 mm × 20 mm, pkg of 5 pieces
Expand
View Pricing
SM75100105
SmartPor, alumina membrane,
thickness 100 μm, pore diameter 75 nm, pore size 125 nm (interpore distance), size 10 mm × 10 mm, pkg of 5 pieces
Expand
View Pricing
SM751001010
SmartPor, alumina membrane,
thickness 100 μm, pore diameter 75 nm, pore size 125 nm (interpore distance), size 10 mm × 10 mm, pkg of 10 pieces
Expand
View Pricing
SM60100105
SmartPor, alumina membrane,
thickness 100 μm, pore diameter 60 nm, pore size 125 nm (interpore distance), size 10 mm × 10 mm, pkg of 5 pieces
Expand
View Pricing
SM75100201
SmartPor, alumina membrane,
thickness 100 μm, pore diameter 75 nm, pore size 125 nm (interpore distance), size 20 mm × 20 mm, pkg of 1 pieces
Expand
View Pricing
SM601001010
SmartPor, alumina membrane,
thickness 100 μm, pore diameter 60 nm, pore size 125 nm (interpore distance), size 10 mm × 10 mm, pkg of 10 pieces
Expand
View Pricing
SM6050205
SmartPor, alumina membrane,
thickness 50 μm, pore diameter 60 nm, pore size 125 nm (interpore distance), size 20 mm × 20 mm, pkg of 5 pieces
Expand
View Pricing
SM60100205
SmartPor, alumina membrane,
thickness 100 μm, pore diameter 60 nm, pore size 125 nm (interpore distance), size 20 mm × 20 mm, pkg of 5 pieces
Expand
View Pricing
SM40100205
SmartPor, alumina membrane,
thickness 100 μm, pore diameter 40 nm, pore size 125 nm (interpore distance), size 20 mm × 20 mm, pkg of 5 pieces
Expand
View Pricing
SM40100201
SmartPor, alumina membrane,
thickness 100 μm, pore diameter 40 nm, pore size 125 nm (interpore distance), size 20 mm × 20 mm, pkg of 1 pieces
Expand
View Pricing
SM40100105
SmartPor, alumina membrane,
thickness 100 μm, pore diameter 40 nm, pore size 125 nm (interpore distance), size 10 mm × 10 mm, pkg of 5 pieces
Expand
View Pricing
Page 1 of 2
Page 1 of 2