Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sleeve stopper septa
Products
Product Category
septa (17)
Brand
Sigma-Aldrich (17)
Color
ID (mm)
OD (mm)
Filter Products
Showing 1-17 of 17
Compare
Product Number
Description
Pricing
Z564702
Sleeve stopper septa,
bottom I.D. × O.D. 7.9 mm × 14 mm, white
Expand
View Pricing
Z564729
Sleeve stopper septa,
bottom I.D. × O.D. 9.3 mm × 15.7 mm, white
Expand
View Pricing
Z564745
Sleeve stopper septa,
bottom I.D. × O.D. 12.7 mm × 23.7 mm, white
Expand
View Pricing
Z564680
Sleeve stopper septa,
bottom I.D. × O.D. 4.2 mm × 10.7 mm, white
Expand
View Pricing
Z565709
Sleeve stopper septa,
bottom I.D. × O.D. 3.4 mm × 6.6 mm, red
Expand
View Pricing
Z565695
Sleeve stopper septa,
bottom I.D. × O.D. 2.4 mm × 5.3 mm, white
Expand
View Pricing
Z564699
Sleeve stopper septa,
bottom I.D. × O.D. 7.9 mm × 14 mm, red
Expand
View Pricing
Z564737
Sleeve stopper septa,
bottom I.D. × O.D. 12.7 mm × 23.7 mm, red
Expand
View Pricing
Z565717
Sleeve stopper septa,
bottom I.D. × O.D. 3.4 mm × 6.6 mm, white
Expand
View Pricing
Z565644
Sleeve stopper septa,
bottom I.D. × O.D. 1.5 mm × 3.9 mm, red
Expand
View Pricing
Z565660
Sleeve stopper septa,
bottom I.D. × O.D. 2.2 mm × 4.5 mm, red
Expand
View Pricing
Z564710
Sleeve stopper septa,
bottom I.D. × O.D. 9.3 mm × 15.7 mm, red
Expand
View Pricing
Z565687
Sleeve stopper septa,
bottom I.D. × O.D. 2.4 mm × 5.3 mm, red
Expand
View Pricing
Z565652
Sleeve stopper septa,
bottom I.D. × O.D. 1.5 mm × 3.9 mm, white
Expand
View Pricing
Z564672
Sleeve stopper septa,
bottom I.D. × O.D. 4.2 mm × 10.7 mm, red
Expand
View Pricing
Z565725
Sleeve stopper septa,
bottom I.D. × O.D. 6.5 mm × 13.3 mm, red
Expand
View Pricing
Z741038
Sleeve stopper septa,
bottom I.D. × O.D. 2.2 mm × 4.5 mm, white
Expand
View Pricing