
Sinalbin potassium salt

Synonym(s):
p-Hydroxybenzylglucosinolate potassium salt, Glucosinalbin potassium salt
Empirical Formula (Hill Notation):
C14H18KNO10S2
CAS Number:
Molecular Weight:
463.52
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing