Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
SimpliPak
®
Purification Cartridge
Products
Product Category
polishers (3)
Brand
Milli-Q (3)
Compatibility
Quality Segment
Pore size (µm)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
SIPK0SIA1
SimpliPak
®
Purification Cartridge,
For Simplicity
®
water purification systems with Elix
®
/ RO / distilled water feed
Expand
View Pricing
SIPK0SIX2
SimpliPak
®
Purification Cartridge,
For Simplicity
®
systems with DI water feed
Expand
View Pricing
SIPK0SIX3
SimpliPak
®
Purification Cartridge,
For Simplicity
®
systems with DI water feed containing high silt
Expand
View Pricing