Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Silver, dispersion
Linear Formula:
Ag
CAS Number:
7440-22-4
Molecular Weight:
107.87
Products
Brand
Sigma-Aldrich (8)
Formula Weight
Physical Form
Quality Segment
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
730785
Silver, dispersion,
nanoparticles, 10 nm particle size (TEM), 0.02 mg/mL in aqueous buffer, contains sodium citrate as stabilizer
Expand
View Pricing
730793
Silver, dispersion,
nanoparticles, 20 nm particle size (TEM), 0.02 mg/mL in aqueous buffer, contains sodium citrate as stabilizer
Expand
View Pricing
730807
Silver, dispersion,
nanoparticles, 40 nm particle size (TEM), 0.02 mg/mL in aqueous buffer, contains sodium citrate as stabilizer
Expand
View Pricing
736465
Silver, dispersion,
nanoparticle, 30-35 wt. % in triethylene glycol monoethyl ether, spec. resistivity 11 μΩ-cm, for printing on plastic films
Expand
View Pricing
730777
Silver, dispersion,
nanoparticles, 100 nm particle size (TEM), 0.02 mg/mL in aqueous buffer, contains sodium citrate as stabilizer
Expand
View Pricing
730815
Silver, dispersion,
nanoparticles, 60 nm particle size (TEM), 0.02 mg/mL in aqueous buffer, contains sodium citrate as stabilizer
Expand
View Pricing
736511
Silver, dispersion,
nanoparticle, 40-50 wt. %, spec. resistivity < 0.001 Ω-cm, for printing on ITO and glass
Expand
View Pricing
736473
Silver, dispersion,
nanoparticle, 30-35 wt. % in triethylene glycol monoethyl ether, spec. resistivity ~ 7 μΩ-cm, for printing on ITO films
Expand
View Pricing