Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Silicon nitride - reaction bonded
Products
Manufacturer
Physical Form
Length (mm)
OD (mm)
Filter Products
Showing 1-13 of 13
Compare
Product Number
Description
Pricing
GF12948958
Silicon nitride - reaction bonded,
tube, outside diameter 5.0 mm, length 100 mm, wall thickness 1.0 mm
Expand
View Pricing
GF73330264
Silicon nitride - reaction bonded,
sheet, thickness 1.0 mm, size 50 × 50 mm
Expand
View Pricing
GF22972158
Silicon nitride - reaction bonded,
rod, 2.0 mm diameter, length 100 mm
Expand
View Pricing
GF85351608
Silicon nitride - reaction bonded,
sheet, thickness 2.0 mm, size 50 × 50 mm
Expand
View Pricing
GF68781167
Silicon nitride - reaction bonded,
sphere, 3 mm diameter, 10 pcs, tolerance +/-2.5um
Expand
View Pricing
GF86194200
Silicon nitride - reaction bonded,
sphere, 6 mm diameter, 10 pcs, tolerance +/-2.5um
Expand
View Pricing
GF18750146
Silicon nitride - reaction bonded,
sheet, thickness 4.0 mm, size 25 × 25 mm
Expand
View Pricing
GF43767603
Silicon nitride - reaction bonded,
sheet, thickness 4.0 mm, size 50 × 50 mm
Expand
View Pricing
GF18619072
Silicon nitride - reaction bonded,
tube, outside diameter 8.0 mm, length 100 mm, wall thickness 1.5 mm
Expand
View Pricing
GF71518989
Silicon nitride - reaction bonded,
rod, 5.0 mm diameter, length 100 mm
Expand
View Pricing
GF39873101
Silicon nitride - reaction bonded,
rod, 10.0 mm diameter, length 150 mm
Expand
View Pricing
GF70950247
Silicon nitride - reaction bonded,
sheet, thickness 4.0 mm, size 100 × 100 mm
Expand
View Pricing
GF25138504
Silicon nitride - reaction bonded,
sphere, 10 mm diameter, 10_pcs, tolerance +/-2.5um
Expand
View Pricing