Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Silicon nitride
Products
Manufacturer
Physical Form
Particle Size (µm)
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
GF44799935
Silicon nitride,
powder, max. particle size 10 micron, weight 100 g, condition alpha si3n4 (85% minimum)
Expand
View Pricing
GF64213185
Silicon nitride,
powder, 10 max. part. size (micron), weight 1000 g, condition alpha si3n4 (85% minimum)
Expand
View Pricing
GF61158393
Silicon nitride,
powder, 10 max. part. size (micron), weight 50 g, condition alpha si3n4 (85% minimum)
Expand
View Pricing
GF85568800
Silicon nitride,
powder, 0.02 max. part. size (micron), weight 100 g, purity 99%
Expand
View Pricing
GF13036070
Silicon nitride,
powder, max. particle size 0.02 micron, weight 200 g, purity 99%
Expand
View Pricing
GF25397848
Silicon nitride,
powder, max. particle size 0.8 micron, weight 200 g, min. particle size 0.1 micron
Expand
View Pricing
GF98913707
Silicon nitride,
powder, 10 max. part. size (micron), weight 500 g, condition alpha si3n4 (85% minimum)
Expand
View Pricing
GF99991300
Silicon nitride,
powder, 10 max. part. size (micron), weight 200 g, condition alpha si3n4 (85% minimum)
Expand
View Pricing