Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sigma-Aldrich
®
standard Ag/AgCl double junction pH combination electrode, glass body
Products
Brand
Sigma-Aldrich (7)
Length (mm)
OD (mm)
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
E4753
Sigma-Aldrich
®
standard Ag/AgCl double junction pH combination electrode, glass body,
Ferrule-pin connector, electrode L 120 mm
Expand
View Pricing
Z113255
Sigma-Aldrich
®
standard Ag/AgCl double junction pH combination electrode, glass body,
Standard pin, electrode L 55 mm , with bulb guard
Expand
View Pricing
Z113298
Sigma-Aldrich
®
standard Ag/AgCl double junction pH combination electrode, glass body,
BNC, electrode L 120 mm
Expand
View Pricing
E5634
Sigma-Aldrich
®
standard Ag/AgCl double junction pH combination electrode, glass body,
BNC Connector, electrode L 110 mm
Expand
View Pricing
E6384
Sigma-Aldrich
®
standard Ag/AgCl double junction pH combination electrode, glass body,
BNC Connector, electrode L 120 mm
Expand
View Pricing
Z113263
Sigma-Aldrich
®
standard Ag/AgCl double junction pH combination electrode, glass body,
BNC, electrode L 55 mm , with bulb guard
Expand
View Pricing
E4878
Sigma-Aldrich
®
standard Ag/AgCl double junction pH combination electrode, glass body,
Ferrule pin connector, electrode L 110 mm
Expand
View Pricing