Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Short thread screw cap, 9 mm thread, with liner
Products
Product Category
septa (5)
caps (1)
Brand
Supelco (5)
Diameter (mm)
OD (mm)
Color
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
SU860092
Short thread screw cap, 9 mm thread, with liner,
blue polypropylene cap (with 6 mm center hole), red PTFE/white silicone septum, pkg of 100 ea
Expand
View Pricing
854161
Short thread screw cap, 9 mm thread, with liner,
blue polypropylene cap, PTFE/red rubber septum, septum diam. × thickness 9 mm × 1.0 mm, pkg of 100 ea
Expand
View Pricing
SU860019
Short thread screw cap, 9 mm thread, with liner,
Expand
View Pricing
854971
Short thread screw cap, 9 mm thread, with liner,
clear cap, PTFE/silicone septum, septum diam. × thickness 9 mm × 1.0 mm, pkg of 1000 ea
Expand
View Pricing
SU860079
Short thread screw cap, 9 mm thread, with liner,
blue polypropylene cap, red PTFE/silicone/red PTFE septum, septum diam. × thickness 9 mm × 1.0 mm, pkg of 100 ea
Expand
View Pricing
854973
Short thread screw cap, 9 mm thread, with liner,
blue cap, PTFE/red rubber septum, thickness 1.0 mm, pkg of 1000 ea
Expand
View Pricing