Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Shigemi 5 mm Symmetrical NMR microtube assembly
Products
Product Category
tubes (16)
Brand
Sigma-Aldrich (5)
Manufacturer
Diameter (mm)
Length (mm)
OD (mm)
Frequency (MHz)
Filter Products
Showing 1-16 of 16
Compare
Product Number
Description
Pricing
Z543349
Shigemi 5 mm Symmetrical NMR microtube assembly,
matched with D
2
O, bottom L 8 mm
Expand
View Pricing
Z529346
Shigemi 5 mm Symmetrical NMR microtube assembly,
matched with CDCl
3
, bottom L 8 mm
Expand
View Pricing
Z693375
Shigemi 5 mm Symmetrical NMR microtube assembly,
matched with CD
3
OD, bottom L 8 mm
Expand
View Pricing
Z529435
Shigemi 5 mm Symmetrical NMR microtube assembly,
matched with D
2
O, bottom L 15 mm
Expand
View Pricing
Z529443
Shigemi 5 mm Symmetrical NMR microtube assembly,
matched with D
2
O, microtube cap only
Expand
View Pricing
Z529370
Shigemi 5 mm Symmetrical NMR microtube assembly,
matched with CDCl
3
, microtube cap
Expand
View Pricing
Z529419
Shigemi 5 mm Symmetrical NMR microtube assembly,
matched with DMSO-d
6
, microtube cap
Expand
View Pricing
Z693405
Shigemi 5 mm Symmetrical NMR microtube assembly,
matched with CD
3
OD, microtube cap
Expand
View Pricing
Z529389
Shigemi 5 mm Symmetrical NMR microtube assembly,
matched with DMSO-d
6
, bottom L 8 mm
Expand
View Pricing
Z529354
Shigemi 5 mm Symmetrical NMR microtube assembly,
matched with CDCl
3
, bottom L 12 mm
Expand
View Pricing
Z693383
Shigemi 5 mm Symmetrical NMR microtube assembly,
matched with CD
3
OD, bottom L 12 mm
Expand
View Pricing
Z529362
Shigemi 5 mm Symmetrical NMR microtube assembly,
matched with CDCl
3
, bottom L 15 mm
Expand
View Pricing
Z529400
Shigemi 5 mm Symmetrical NMR microtube assembly,
matched with DMSO-d
6
, bottom L 15 mm
Expand
View Pricing
Z693391
Shigemi 5 mm Symmetrical NMR microtube assembly,
matched with CD
3
OD, bottom L 15 mm
Expand
View Pricing
Z529397
Shigemi 5 mm Symmetrical NMR microtube assembly,
matched with DMSO-d
6
, bottom L 12 mm
Expand
View Pricing
Z529427
Shigemi 5 mm Symmetrical NMR microtube assembly,
matched with D
2
O, bottom L 12 mm
Expand
View Pricing