
Sevoflurane Related Compound B

Synonym(s):
1,1,1,3,3,3-Hexafluoro-2-methoxypropane, Hexafluoroisopropyl methyl ether
Empirical Formula (Hill Notation):
C4H4F6O
CAS Number:
Molecular Weight:
182.06
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2