Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Screw cap, phenolic, with open center
Products
Product Category
caps (5)
Brand
Supelco (5)
Diameter (mm)
Closure Type
Color
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
27187
Screw cap, phenolic, with open center,
black phenolic hole cap, for use with 20 or 40 mL vial with 24-400 thread, pkg of 100 ea
Expand
View Pricing
27176
Screw cap, phenolic, with open center,
black phenolic hole cap, for use with 22 mL vial with 20-400 thread, pkg of 100 ea
Expand
View Pricing
27154
Screw cap, phenolic, with open center,
black phenolic hole cap, for use with 7 mL vial with 15-425 thread, pkg of 100 ea
Expand
View Pricing
27120-U
Screw cap, phenolic, with open center,
black phenolic cap, for use with 4 mL vial with 13-425 thread, pkg of 100 ea
Expand
View Pricing
23237
Screw cap, phenolic, with open center,
black phenolic cap (no septum), thread for 24-400, pkg of 144 ea
Expand
View Pricing