Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Screw cap (open-top), 9 mm thread, with liner
Products
Product Category
septa (7)
caps (1)
Brand
Supelco (5)
Height (mm)
ID (mm)
OD (mm)
Closure Type
Color
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
29047-U
Screw cap (open-top), 9 mm thread, with liner,
blue polypropylene cap, PTFE/silicone septum, pkg of 100 ea
Expand
View Pricing
29049-U
Screw cap (open-top), 9 mm thread, with liner,
black polypropylene cap, PTFE/silicone septum (with slit), pkg of 100 ea
Expand
View Pricing
29051-U
Screw cap (open-top), 9 mm thread, with liner,
blue polypropylene cap, PTFE/silicone septum (with slit), pkg of 100 ea
Expand
View Pricing
27326
Screw cap (open-top), 9 mm thread, with liner,
clear cap, PTFE/silicone septum, pkg of 100 ea
Expand
View Pricing
27560-U
Screw cap (open-top), 9 mm thread, with liner,
clear cap, PTFE/silicone/PTFE septum, pkg of 1000 ea
Expand
View Pricing
27558
Screw cap (open-top), 9 mm thread, with liner,
clear cap, PTFE/silicone septum, pkg of 1000 ea
Expand
View Pricing
507415
Screw cap (open-top), 9 mm thread, with liner,
red polypropylene cap, tan PTFE/silicone septum, pkg of 100 ea
Expand
View Pricing
SU860143
Screw cap (open-top), 9 mm thread, with liner,
PTFE/silicone (with slit)
Expand
View Pricing