Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Scissor-handle forceps
Products
Product Category
forceps (9)
Brand
Sigma-Aldrich (2)
Length (in)
Package Size
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
Z168831
Scissor-handle forceps,
Kelly hemostatic, straight, L 5 1/2 in.
Expand
View Pricing
Z168874
Scissor-handle forceps,
curved, Kelly hemostatic, L 6 1/4 in.
Expand
View Pricing
Z168815
Scissor-handle forceps,
Halstead mosquito, straight, L 5 in.
Expand
View Pricing
Z168823
Scissor-handle forceps,
Halstead mosquito, curved, L 5 in.
Expand
View Pricing
Z168858
Scissor-handle forceps,
curved, Kelly hemostatic, L 5 1/2 in.
Expand
View Pricing
Z509140
Scissor-handle forceps,
straight, Occulating, L 7 in.
Expand
View Pricing
Z168866
Scissor-handle forceps,
straight, Kelly hemostatic, L 6 1/4 in.
Expand
View Pricing
Z509132
Scissor-handle forceps,
Occulating, straight, L 5 in.
Expand
View Pricing
Z168807
Scissor-handle forceps,
Specimen, straight, L 10 1/4 in.
Expand
View Pricing