Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Scienceware
®
ROTO-FLO
™
flow indicator
Products
Brand
Sigma-Aldrich (1)
Color
Manufacturer
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
Z408468
Scienceware
®
ROTO-FLO
™
flow indicator,
fits tubing, 5/16 - 1/2in., green wheel
Expand
View Pricing
Z408441
Scienceware
®
ROTO-FLO
™
flow indicator,
fits tubing, 1/4 ‑ 5/16 in., red wheel
Expand
View Pricing
Z408433
Scienceware
®
ROTO-FLO
™
flow indicator,
fits tubing, 3/16 ‑ 1/4 in., blue wheel
Expand
View Pricing