Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Scienceware
®
Lab Companion
™
round vacuum desiccator
Products
Product Category
desiccators (6)
Manufacturer
Volume (ml)
Color
Package Size
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
BAF424002021
Scienceware
®
Lab Companion
™
round vacuum desiccator,
volume 6 L, clear polycarbonate, pack of 1 ea
Expand
View Pricing
BAF424002121
Scienceware
®
Lab Companion
™
round vacuum desiccator,
volume 10 L, clear polycarbonate, pack of 1 ea
Expand
View Pricing
BAF424002221
Scienceware
®
Lab Companion
™
round vacuum desiccator,
volume 20 L, clear polycarbonate, pack of 1 ea
Expand
View Pricing
BAF424002141
Scienceware
®
Lab Companion
™
round vacuum desiccator,
volume 10 L, amber polycarbonate (UV)
Expand
View Pricing
BAF424002041
Scienceware
®
Lab Companion
™
round vacuum desiccator,
volume 6 L, amber polycarbonate (UV), pack of 1 ea
Expand
View Pricing
BAF424002241
Scienceware
®
Lab Companion
™
round vacuum desiccator,
volume 20 L, amber polycarbonate (UV), pack of 1 ea
Expand
View Pricing