Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sartorius filter pipette tips
Products
Product Category
pipette tips (7)
Manufacturer
Binding Type
Volume Range (µL)
Color
Package Size
Sterility
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
Z757802
Sartorius filter pipette tips,
volume range 5-300 μL, sterile, pack of 960 ea (10 racks of 96tips)
Expand
View Pricing
Z757799
Sartorius filter pipette tips,
volume range 5-200 μL, sterile, pack of 960 ea (10 racks of 96 tips)
Expand
View Pricing
Z757780
Sartorius filter pipette tips,
volume range 2-120 μL, sterile, pack of 960 ea (10 racks of 96 tips)
Expand
View Pricing
Z757810
Sartorius filter pipette tips,
volume range 50-1000 μL, sterile, pack of 960 ea (10 racks of 96 tips)
Expand
View Pricing
Z757764
Sartorius filter pipette tips,
volume range 0.1-10 μL, sterile, pack of 960 ea (10 racks of 96 tips)
Expand
View Pricing
Z757772
Sartorius filter pipette tips,
volume range 0.5-20 μL, sterile, pack of 960 ea (10 racks of 96 tips)
Expand
View Pricing
Z757829
Sartorius filter pipette tips,
volume range 50-1200 μL, sterile, pack of 960 ea (10 racks of 96 tips)
Expand
View Pricing