Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Rubbermaid
®
heavy-duty lab carts
Products
Brand
Sigma-Aldrich (3)
Color
Length (in)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
Z208175
Rubbermaid
®
heavy-duty lab carts,
black
Expand
View Pricing
Z208221
Rubbermaid
®
heavy-duty lab carts,
Model B, H × L × W 32 1/2 in. × 36 in. × 24 in., beige, cart total capacity 400 lb. , 200 lb. per shelf
Expand
View Pricing
Z208213
Rubbermaid
®
heavy-duty lab carts,
Model B, H × W × L 33 1/4 in. × 25 7/8 in. × 45 1/4 in., black, cart total capacity 500 lb. , 250 lb. per shelf
Expand
View Pricing