
Rocuronium for peak identification

Synonym(s):
Rocuronium bromide
Empirical Formula (Hill Notation):
C32H53BrN2O4
CAS Number:
Molecular Weight:
609.68
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing