Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Residual Solvent Class 2 -
N
,
N
-Dimethylacetamide
Synonym(s):
N,N
-Dimethylacetamide solution
Empirical Formula (Hill Notation):
C
4
H
9
NO
CAS Number:
127-19-5
Molecular Weight:
87.12
EC Number:
204-826-4
Products
Formula Weight
Manufacturer
Format
Application
Reference Material Type
Special Grade
pharmaceutical primary standard (1)
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
1601485
Residual Solvent Class 2 -
N
,
N
-Dimethylacetamide,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing