Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Real Crystal
®
IR sample cards
Products
Brand
Sigma-Aldrich (9)
Diameter (mm)
Technique
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
Z527084
Real Crystal
®
IR sample cards,
KBr, aperture diam. 9.5 mm
Expand
View Pricing
Z527076
Real Crystal
®
IR sample cards,
NaCl, aperture diam. 9.5 mm
Expand
View Pricing
Z527149
Real Crystal
®
IR sample cards,
KBr, aperture diam. 19 mm
Expand
View Pricing
Z527114
Real Crystal
®
IR sample cards,
KBr, aperture diam. 15 mm
Expand
View Pricing
Z527173
Real Crystal
®
IR sample cards,
Crystal cover slips for Z527084
Expand
View Pricing
Z527165
Real Crystal
®
IR sample cards,
Crystal cover slips for Z527076
Expand
View Pricing
Z527106
Real Crystal
®
IR sample cards,
NaCl, aperture diam. 15 mm
Expand
View Pricing
Z527130
Real Crystal
®
IR sample cards,
NaCl, aperture diam. 19 mm
Expand
View Pricing
Z527122
Real Crystal
®
IR sample cards,
KCl, aperture diam. 15 mm
Expand
View Pricing