Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Reagent Reservoir
Products
Manufacturer
Capacity (mL)
Package Size
Sterility
Sterilization Method
Filter Products
Showing 1-17 of 17
Compare
Product Number
Description
Pricing
HS120637
Reagent Reservoir,
capacity 5 mL, polystyrene, sterile, pack of 50 (individually wrapped)
Expand
View Pricing
HS120639
Reagent Reservoir,
capacity 10 mL, polystyrene, sterile; electron beam irradiated, pack of 100 (individually wrapped)
Expand
View Pricing
HS120641
Reagent Reservoir,
capacity 25 mL, polystyrene, sterile; electron beam irradiated, pack of 100 (individually wrapped)
Expand
View Pricing
HS120638
Reagent Reservoir,
capacity 10 mL, polystyrene, non-sterile, pack of 100 (bulk packed)
Expand
View Pricing
HS120642
Reagent Reservoir,
reservoir capacity 25 mL, polystyrene reservoir, non-sterile, pack of 100 (bulk packed)
Expand
View Pricing
HS120640
Reagent Reservoir,
capacity 25 mL, polystyrene, sterile; electron beam irradiated, pack of 200 (packed as 5 ea)
Expand
View Pricing
HS120648
Reagent Reservoir,
capacity 100 mL, polystyrene, sterile; electron beam irradiated, pack of 100 (individually wrapped)
Expand
View Pricing
HS120644
Reagent Reservoir,
capacity 50 mL, polystyrene, sterile; electron beam irradiated, pack of 100 (packed as 5 ea)
Expand
View Pricing
HS120645
Reagent Reservoir,
capacity 50 mL, polystyrene, non-sterile, pack of 100 (bulk packed)
Expand
View Pricing
HS120649
Reagent Reservoir,
capacity 100 mL, polystyrene, non-sterile, pack of 100 (bulk packed)
Expand
View Pricing
HS120643
Reagent Reservoir,
capacity 50 mL, polystyrene, sterile; electron beam irradiated, pack of 200 (packed as 5 ea)
Expand
View Pricing
HS120690
Reagent Reservoir,
capacity 50 mL, PVC, non-sterile, pack of 100 (bulk packed)
Expand
View Pricing
HS120646
Reagent Reservoir,
capacity 50 mL, PVC, sterile; electron beam irradiated, pack of 100 (packed as 5 ea)
Expand
View Pricing
HS120652
Reagent Reservoir,
capacity 25 mL, polystyrene, sterile; electron beam irradiated, pack of 100 (individually wrapped)
Expand
View Pricing
HS120647
Reagent Reservoir,
capacity 100 mL, polystyrene, sterile; electron beam irradiated, pack of 200 (packed as 5 ea)
Expand
View Pricing
HS120651
Reagent Reservoir,
capacity 25 mL, polystyrene, sterile; electron beam irradiated, pack of 200 (packed as 5 ea)
Expand
View Pricing
HS120650
Reagent Reservoir,
capacity 100 mL, PVC, non-sterile, pack of 100 (bulk packed)
Expand
View Pricing