Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Rare earth extra power stir bars
Products
Product Category
stir bars (9)
Manufacturer
Diameter (mm)
Length (mm)
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
Z671657
Rare earth extra power stir bars,
Elliptical, diam. 6 mm, L 10 mm
Expand
View Pricing
Z671630
Rare earth extra power stir bars,
oval, diam. 10 mm, L 20 mm
Expand
View Pricing
Z671622
Rare earth extra power stir bars,
oval, diam. 5 mm, L 10 mm
Expand
View Pricing
Z671649
Rare earth extra power stir bars,
oval, diam. 12 mm, L 25 mm
Expand
View Pricing
Z671665
Rare earth extra power stir bars,
Elliptical, diam. 10 mm
Expand
View Pricing
Z671673
Rare earth extra power stir bars,
Elliptical, diam. 14 mm, L 25 mm
Expand
View Pricing
Z671592
Rare earth extra power stir bars,
Octahedral, diam. 8 mm, L 13 mm
Expand
View Pricing
Z671614
Rare earth extra power stir bars,
octagonal (with pivot ring), diam. 8 mm, L 38 mm
Expand
View Pricing
Z671606
Rare earth extra power stir bars,
octagonal (with pivot ring), diam. 8 mm, L 25 mm
Expand
View Pricing