Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Quickfit
®
conical flasks
Products
Product Category
flasks (17)
Manufacturer
Capacity (mL)
Cap
Closure Type
Package Size
Sterility
Filter Products
Showing 1-17 of 17
Compare
Product Number
Description
Pricing
Z308714
Quickfit
®
conical flasks,
capacity 50 mL, joint: ST/NS 24/29
Expand
View Pricing
Z308838
Quickfit
®
conical flasks,
capacity 250 mL, joint: ST/NS 24/29
Expand
View Pricing
Z308862
Quickfit
®
conical flasks,
capacity 500 mL, joint: ST/NS 19/26
Expand
View Pricing
Z308870
Quickfit
®
conical flasks,
capacity 500 mL, joint: ST/NS 24/29
Expand
View Pricing
Z308749
Quickfit
®
conical flasks,
capacity 100 mL, joint: ST/NS 24/29
Expand
View Pricing
Z308706
Quickfit
®
conical flasks,
joint: ST/NS 19/26
Expand
View Pricing
Z308900
Quickfit
®
conical flasks,
capacity 1000 mL, joint: ST/NS 24/29
Expand
View Pricing
Z308722
Quickfit
®
conical flasks,
capacity 100 mL, joint: ST/NS 14/23
Expand
View Pricing
Z308889
Quickfit
®
conical flasks,
capacity 500 mL, joint: ST/NS 29/32
Expand
View Pricing
Z308846
Quickfit
®
conical flasks,
capacity 250 mL, joint: ST/NS 29/32
Expand
View Pricing
Z308730
Quickfit
®
conical flasks,
capacity 100 mL, joint: ST/NS 19/26
Expand
View Pricing
Z308757
Quickfit
®
conical flasks,
capacity 100 mL, joint: ST/NS 29/32
Expand
View Pricing
Z308668
Quickfit
®
conical flasks,
capacity 25 mL, joint: ST/NS 14/23
Expand
View Pricing
Z308692
Quickfit
®
conical flasks,
capacity 50 mL, joint: ST/NS 14/23
Expand
View Pricing
Z308811
Quickfit
®
conical flasks,
capacity 250 mL, joint: ST/NS 19/26
Expand
View Pricing
Z308781
Quickfit
®
conical flasks,
capacity 150 mL, joint: ST/NS 24/29
Expand
View Pricing
Z308803
Quickfit
®
conical flasks,
capacity 150 mL, joint: ST/NS 29/32
Expand
View Pricing